Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSIR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005565587.2 | XP_005565644.1 | 1 - 2 | 257 | 62 | |
XM_015456211.1 | XP_015311697.1 | 1 - 2 | 259 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Inventoried Probe spans exons |