Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: IL25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005560861.1 | XP_005560918.1 | 1 - 2 | 273 | 88 | |
XM_005560862.2 | XP_005560919.1 | 2 - 3 | 1275 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Inventoried Probe spans exons |