Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TARBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005571030.2 | XP_005571087.1 | 6 - 7 | 2014 | 154 | |
XM_005571031.2 | XP_005571088.1 | 6 - 7 | 700 | 154 | ||
XM_005571032.2 | XP_005571089.1 | 6 - 7 | 675 | 154 | ||
XM_005571033.2 | XP_005571090.1 | 6 - 7 | 684 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |