Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102140229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005590834.2 | XP_005590891.1 | 4 - 5 | 1108 | 75 | |
GenBank mRNA | EU341821.1 | - | 4 - 5 | 645 | 75 | |
EU341822.1 | - | 4 - 5 | 645 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |