Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102135631 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015439652.1 | XP_015295138.1 | 8 | 1172 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |