Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc11a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146161.1 | NP_001139633.1 | 7 - 8 | 741 | 95 | |
NM_008732.2 | NP_032758.2 | 7 - 8 | 741 | 95 | ||
GenBank mRNA | AF029758.1 | - | 7 - 8 | 676 | 95 | |
AK049856.1 | - | 7 - 8 | 689 | 95 | ||
AK083478.1 | - | 7 - 8 | 687 | 95 | ||
AK148276.1 | - | 7 - 8 | 689 | 95 | ||
AK213433.1 | - | 3 - 4 | 223 | 95 | ||
BC019137.1 | - | 7 - 8 | 715 | 5040976 | 95 | |
L33415.1 | - | 7 - 8 | 669 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |