Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008795.2 | NP_032821.1 | 12 - 13 | 1412 | 103 | |
GenBank mRNA | AK004998.1 | - | 12 - 13 | 1356 | 103 | |
AK171569.1 | - | 12 - 13 | 1193 | 103 | ||
BC119617.1 | - | 13 - 14 | 1288 | 40061728 | 103 | |
BC119618.1 | - | 7 - 8 | 679 | 103 | ||
X69026.1 | - | 12 - 13 | 1429 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |