Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3cd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029837.2 | NP_001025008.2 | 7 - 8 | 1206 | 67 | |
NM_001164049.1 | NP_001157521.1 | 7 - 8 | 1233 | 67 | ||
NM_001164050.1 | NP_001157522.1 | 7 - 8 | 1233 | 67 | ||
NM_001164051.1 | NP_001157523.1 | 7 - 8 | 1257 | 67 | ||
NM_001164052.1 | NP_001157524.1 | 5 - 6 | 960 | 67 | ||
NM_008840.3 | NP_032866.2 | 8 - 9 | 1408 | 67 | ||
GenBank mRNA | AK040867.1 | - | 7 - 8 | 1168 | 67 | |
AK149935.1 | - | 8 - 9 | 1307 | 67 | ||
AK150649.1 | - | 7 - 8 | 1134 | 67 | ||
AK151482.1 | - | 7 - 8 | 1134 | 67 | ||
AK152661.1 | - | 7 - 8 | 1134 | 67 | ||
AK153304.1 | - | 7 - 8 | 1168 | 67 | ||
AK155835.1 | - | 7 - 8 | 1119 | 67 | ||
AK171025.1 | - | 7 - 8 | 1216 | 67 | ||
AK172222.1 | - | 7 - 8 | 1110 | 67 | ||
BC035203.1 | - | 7 - 8 | 1223 | 4192906 | 67 | |
CT010349.1 | - | 5 - 6 | 928 | 67 | ||
U86587.1 | - | 5 - 6 | 928 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |