Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pthlh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008970.3 | NP_032996.2 | 1 - 2 | 508 | 65 | |
GenBank mRNA | AY220497.1 | - | 1 - 2 | 269 | 65 | |
BC058187.1 | - | 1 - 2 | 313 | 5346064 | 65 | |
CB194603.1 | - | 1 - 2 | 590 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |