Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009897.2 | NP_034027.1 | 5 - 6 | 964 | 70 | |
GenBank mRNA | AK005076.1 | - | 5 - 6 | 963 | 70 | |
AK131953.1 | - | 5 - 6 | 968 | 70 | ||
BC025976.1 | - | 6 - 7 | 879 | 4216758 | 70 | |
Z13968.1 | - | 5 - 6 | 964 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |