Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ilk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010562.2 | NP_034692.2 | 1 - 2 | 86 | 79 | |
GenBank mRNA | U94479.1 | - | 1 - 2 | 69 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Inventoried Probe spans exons |