Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltbr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010736.3 | NP_034866.1 | 7 - 8 | 948 | 55 | |
GenBank mRNA | AK078859.1 | - | 3 - 4 | 207 | 55 | |
AK146127.1 | - | 7 - 8 | 931 | 55 | ||
BC138590.1 | - | 7 - 8 | 811 | 8861610 | 55 | |
BC138591.1 | - | 7 - 8 | 811 | 8861611 | 55 | |
L38423.1 | - | 7 - 8 | 948 | 55 | ||
U29173.1 | - | 7 - 8 | 932 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |