Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038839.2 | NP_001033928.1 | 4 - 5 | 599 | 74 | |
NM_001038845.2 | NP_001033934.1 | 4 - 5 | 599 | 74 | ||
NM_001284402.1 | NP_001271331.1 | 4 - 5 | 427 | 74 | ||
NM_011027.3 | NP_035157.2 | 4 - 5 | 599 | 74 | ||
GenBank mRNA | AJ009823.1 | - | 4 - 5 | 447 | 74 | |
AJ489296.1 | - | 4 - 5 | 436 | 74 | ||
AJ489297.1 | - | 4 - 5 | 436 | 74 | ||
AK144585.1 | - | 4 - 5 | 600 | 74 | ||
AK145050.1 | - | 4 - 5 | 454 | 74 | ||
AK145211.1 | - | 4 - 5 | 463 | 74 | ||
AK171132.1 | - | 4 - 5 | 571 | 74 | ||
BC128303.1 | - | 4 - 5 | 443 | 74 | ||
BC128304.1 | - | 4 - 5 | 443 | 74 | ||
BC141120.1 | - | 4 - 5 | 466 | 9055732 | 74 | |
FJ436444.1 | - | 4 - 5 | 427 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |