Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sema3e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011348.2 | NP_035478.2 | 13 - 14 | 2136 | 99 | |
GenBank mRNA | AF034744.1 | - | 13 - 14 | 1871 | 99 | |
AK129116.1 | - | 8 - 9 | 901 | 99 | ||
AK142340.1 | - | 13 - 14 | 1963 | 99 | ||
AK159214.1 | - | 13 - 14 | 1927 | 99 | ||
BC057956.1 | - | 13 - 14 | 2034 | 5357516 | 99 | |
Z80941.1 | - | 13 - 14 | 1502 | 99 | ||
Z93947.1 | - | 13 - 14 | 2114 | 99 | ||
Z93948.1 | - | 13 - 14 | 2114 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |