Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1h3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177730.1 | NP_001171201.1 | 3 - 4 | 420 | 115 | |
NM_013839.4 | NP_038867.2 | 3 - 4 | 573 | 115 | ||
GenBank mRNA | AF085745.1 | - | 3 - 4 | 315 | 115 | |
AJ132601.1 | - | 3 - 4 | 351 | 115 | ||
AK143904.1 | - | 3 - 4 | 354 | 115 | ||
AK159250.1 | - | 3 - 4 | 574 | 115 | ||
AY195870.1 | - | 3 - 4 | 420 | 115 | ||
AY195871.1 | - | 3 - 4 | 420 | 115 | ||
BC012646.1 | - | 3 - 4 | 361 | 4163109 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |