Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031197.2 | NP_112474.2 | 4 - 5 | 611 | 83 | |
GenBank mRNA | AK005068.1 | - | 4 - 5 | 612 | 83 | |
AK165430.1 | - | 4 - 5 | 546 | 83 | ||
BC034675.1 | - | 4 - 5 | 530 | 4236331 | 83 | |
X15684.1 | - | 4 - 5 | 560 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |