Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011203.2 | NP_035333.2 | 12 - 13 | 1074 | 81 | |
GenBank mRNA | AK154107.1 | - | 12 - 13 | 1166 | 81 | |
BC051980.1 | - | 12 - 13 | 1074 | 6402545 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |