Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011519.2 | NP_035649.1 | 2 - 3 | 387 | 131 | |
GenBank mRNA | AK132236.1 | - | 2 - 3 | 429 | 131 | |
AK132491.1 | - | 2 - 3 | 370 | 131 | ||
AK143529.1 | - | 2 - 3 | 399 | 131 | ||
AK162092.1 | - | 2 - 3 | 372 | 131 | ||
AK168091.1 | - | 2 - 3 | 276 | 131 | ||
BC010560.1 | - | 2 - 3 | 409 | 3501359 | 131 | |
X15487.1 | - | 2 - 3 | 386 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |