Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ngef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111314.1 | NP_001104784.1 | 8 - 9 | 1525 | 61 | |
NM_019867.2 | NP_063920.2 | 6 - 7 | 1162 | 61 | ||
GenBank mRNA | AJ238898.1 | - | 6 - 7 | 1143 | 61 | |
AK138433.1 | - | 6 - 7 | 1131 | 61 | ||
AY038025.1 | - | 6 - 7 | 1137 | 61 | ||
BC039279.1 | - | 8 - 9 | 1525 | 4973826 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |