Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poll | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020032.2 | NP_064416.1 | 3 - 4 | 489 | 67 | |
GenBank mRNA | AF176099.1 | - | 2 - 3 | 430 | 67 | |
AJ131889.1 | - | 3 - 4 | 489 | 67 | ||
AK133087.1 | - | 3 - 4 | 456 | 67 | ||
BC004767.1 | - | 2 - 3 | 439 | 3156259 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |