Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028347.2 | NP_082623.1 | 5 - 6 | 852 | 125 | |
GenBank mRNA | AB079069.1 | - | 5 - 6 | 852 | 125 | |
AK013322.1 | - | 6 - 7 | 923 | 125 | ||
AK155405.1 | - | 7 - 8 | 728 | 125 | ||
BC034873.1 | - | 6 - 7 | 1097 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |