Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053204.2 | NP_444434.2 | 4 - 5 | 1658 | 99 | |
NM_178085.3 | NP_835186.1 | 3 - 4 | 929 | 99 | ||
NR_104484.1 | 4 - 5 | 1658 | 99 | |||
GenBank mRNA | AF340028.1 | - | 4 - 5 | 1642 | 99 | |
AF340029.1 | - | 4 - 5 | 1642 | 99 | ||
AK035989.1 | - | 3 - 4 | 1472 | 99 | ||
AK048990.1 | - | 3 - 4 | 882 | 99 | ||
AY316692.1 | - | 2 - 3 | 1090 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |