Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Casc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138660.2 | NP_619601.2 | 1 - 2 | 281 | 76 | |
GenBank mRNA | AF526276.1 | - | 1 - 2 | 255 | 76 | |
AJ292072.1 | - | 495 | 76 | |||
AK139608.1 | - | 1 - 2 | 320 | 76 | ||
BC060672.1 | - | 1 - 2 | 109 | 76 | ||
BC141296.1 | - | 1 - 2 | 394 | 9055908 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |