Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146239.2 | NP_666351.2 | 9 - 10 | 1226 | 64 | |
GenBank mRNA | BC031778.1 | - | 773 | 4501115 | 64 | |
BC049904.1 | - | 773 | 4501115 | 64 | ||
BC064815.1 | - | 9 - 10 | 1211 | 6516981 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |