Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubqln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026842.4 | NP_081118.4 | 4 - 5 | 901 | 111 | |
NM_152234.2 | NP_689420.1 | 4 - 5 | 901 | 111 | ||
GenBank mRNA | AK048534.1 | - | 4 - 5 | 822 | 111 | |
AK049298.1 | - | 4 - 5 | 656 | 111 | ||
AK165456.1 | - | 4 - 5 | 866 | 111 | ||
AK172695.1 | - | 4 - 5 | 870 | 111 | ||
BC010213.1 | - | 1 - 2 | 244 | 111 | ||
BC026847.1 | - | 4 - 5 | 884 | 4015641 | 111 | |
BC028857.1 | - | 4 - 5 | 840 | 4507759 | 111 | |
BC051098.1 | - | 3 - 4 | 529 | 111 | ||
BC080667.1 | - | 4 - 5 | 862 | 4935292 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |