Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK010218.1 | - | 2 - 3 | 340 | 60 | |
AK038631.1 | - | 2 - 3 | 388 | 60 | ||
AK156731.1 | - | 2 - 3 | 314 | 60 | ||
AY176058.1 | - | 2 - 3 | 347 | 60 | ||
AY176059.1 | - | 2 - 3 | 378 | 60 | ||
BC021938.1 | - | 2 - 3 | 345 | 5136809 | 60 | |
U19799.1 | - | 2 - 3 | 359 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |