Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164477.1 | NP_001157949.1 | 8 - 9 | 1948 | 65 | |
NM_027835.3 | NP_082111.2 | 9 - 10 | 2095 | 65 | ||
GenBank mRNA | AF374384.1 | - | 9 - 10 | 2004 | 65 | |
AK037057.1 | - | 9 - 10 | 1993 | 65 | ||
AK040519.1 | - | 9 - 10 | 2052 | 65 | ||
AK153018.1 | - | 9 - 10 | 2002 | 65 | ||
AY075132.1 | - | 9 - 10 | 1770 | 65 | ||
BC004031.1 | - | 2 - 3 | 237 | 65 | ||
BC025508.1 | - | 4 - 5 | 539 | 65 | ||
BC080200.1 | - | 8 - 9 | 1853 | 3597815 | 65 | |
EF113547.1 | - | 9 - 10 | 1784 | 65 | ||
EF113548.1 | - | 9 - 10 | 1784 | 65 | ||
EU084750.1 | - | 9 - 10 | 1784 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |