Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Kremen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032396.3 | NP_115772.2 | 2 - 3 | 340 | 92 | |
GenBank mRNA | AB059617.1 | - | 2 - 3 | 303 | 92 | |
AK141321.1 | - | 2 - 3 | 352 | 92 | ||
BC049771.1 | - | 2 - 3 | 264 | 92 | ||
BC082546.1 | - | 2 - 3 | 340 | 6855459 | 92 | |
BC138464.1 | - | 2 - 3 | 348 | 8861484 | 92 | |
BC138465.1 | - | 2 - 3 | 348 | 8861485 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus 1700 Probe spans exons |