Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Commd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB076722.1 | - | 1 - 2 | 676 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Inventoried Probe spans exons |