Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271353.1 | NP_001258282.1 | 9 - 10 | 1191 | 65 | |
NM_144855.3 | NP_659104.1 | 9 - 10 | 1194 | 65 | ||
NM_178224.3 | NP_835742.1 | 9 - 10 | 1194 | 65 | ||
GenBank mRNA | AK077669.1 | - | 9 - 10 | 1155 | 65 | |
AK085692.1 | - | 9 - 10 | 1158 | 65 | ||
BC013472.1 | - | 9 - 10 | 1120 | 4221477 | 65 | |
BC013480.1 | - | 9 - 10 | 1130 | 4239757 | 65 | |
BC022144.1 | - | 9 - 10 | 1155 | 65 | ||
BC026595.1 | - | 9 - 10 | 1127 | 4974777 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |