Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145139.2 | NP_660121.2 | 11 - 12 | 1613 | 67 | |
GenBank mRNA | AJ310346.1 | - | 11 - 12 | 1623 | 67 | |
AK032936.1 | - | 11 - 12 | 1601 | 67 | ||
AK075584.1 | - | 11 - 12 | 1619 | 67 | ||
AK150895.1 | - | 11 - 12 | 1608 | 67 | ||
AK151175.1 | - | 11 - 12 | 1608 | 67 | ||
AK167640.1 | - | 11 - 12 | 1608 | 67 | ||
AK168009.1 | - | 11 - 12 | 1611 | 67 | ||
AK168295.1 | - | 11 - 12 | 1610 | 67 | ||
BC021351.1 | - | 2281 | 67 | |||
BC024463.1 | - | 11 - 12 | 1613 | 4975621 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |