Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153056.2 | NP_694696.2 | 7 - 8 | 846 | 91 | |
GenBank mRNA | AB221687.1 | - | 3 - 4 | 366 | 91 | |
AY251540.1 | - | 7 - 8 | 822 | 91 | ||
BC026403.1 | - | 7 - 8 | 811 | 4209498 | 91 | |
BC026637.1 | - | 6 - 7 | 887 | 91 | ||
BC026650.1 | - | 7 - 8 | 814 | 4987746 | 91 | |
BC040759.1 | - | 7 - 8 | 811 | 4209498 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |