Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032544.3 | NP_115933.2 | 4 - 5 | 1271 | 97 | |
GenBank mRNA | AF361482.2 | - | 4 - 5 | 1271 | 97 | |
AK046330.1 | - | 4 - 5 | 647 | 97 | ||
BC138560.1 | - | 4 - 5 | 680 | 8861580 | 97 | |
BC145333.1 | - | 4 - 5 | 676 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |