Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030558.2 | NP_085035.1 | 1 - 2 | 147 | 80 | |
GenBank mRNA | AF231122.1 | - | 1 - 2 | 99 | 80 | |
AK131988.1 | - | 1 - 2 | 175 | 80 | ||
BC019975.1 | - | 1 - 2 | 147 | 4223351 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |