Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033573.2 | NP_291051.1 | 3 - 4 | 1304 | 63 | |
GenBank mRNA | AF273750.1 | - | 3 - 4 | 1325 | 63 | |
AK078394.1 | - | 3 - 4 | 1324 | 63 | ||
BC027831.1 | - | 3 - 4 | 1337 | 63 | ||
BC052072.1 | - | 3 - 4 | 1304 | 5717482 | 63 | |
BC082784.1 | - | 3 - 4 | 1322 | 4191155 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |