Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Smarca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290708.1 | NP_001277637.1 | 8 - 9 | 1261 | 109 | |
NM_053123.4 | NP_444353.3 | 8 - 9 | 1261 | 109 | ||
GenBank mRNA | AF325920.1 | - | 8 - 9 | 1268 | 109 | |
AK030741.1 | - | 8 - 9 | 1228 | 109 | ||
AK032978.1 | - | 8 - 9 | 1229 | 109 | ||
AK052092.1 | - | 8 - 9 | 1217 | 109 | ||
BC016071.1 | - | 8 - 9 | 1238 | 109 | ||
BC057115.1 | - | 8 - 9 | 1261 | 6406330 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |