Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ap2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077264.1 | NP_001070732.1 | 14 - 15 | 2099 | 91 | |
NM_007458.2 | NP_031484.1 | 14 - 15 | 2099 | 91 | ||
GenBank mRNA | AK135424.1 | - | 14 - 15 | 2028 | 91 | |
AK155515.1 | - | 14 - 15 | 2036 | 91 | ||
BC031433.1 | - | 14 - 15 | 2049 | 4481643 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |