Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007531.2 | NP_031557.2 | 5 - 6 | 754 | 93 | |
GenBank mRNA | AK011107.1 | - | 5 - 6 | 721 | 93 | |
AK090088.1 | - | 5 - 6 | 94 | |||
AK132052.1 | - | 5 - 6 | 716 | 93 | ||
AK132058.1 | - | 5 - 6 | 743 | 93 | ||
AK151952.1 | - | 5 - 6 | 748 | 93 | ||
AK152375.1 | - | 4 - 5 | 1403 | 93 | ||
AK153368.1 | - | 4 - 5 | 1403 | 93 | ||
AY211613.1 | - | 5 - 6 | 716 | 93 | ||
AY319418.1 | - | 5 - 6 | 606 | 93 | ||
BC145875.1 | - | 5 - 6 | 706 | 40131178 | 93 | |
X78683.1 | - | 5 - 6 | 710 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |