Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010925.2 | NP_035055.2 | 4 - 5 | 446 | 83 | |
GenBank mRNA | AF312394.1 | - | 3 - 4 | 209 | 83 | |
BC022910.1 | - | 3 - 4 | 312 | 4165783 | 83 | |
BC065993.1 | - | 4 - 5 | 408 | 5684672 | 83 | |
BC065994.1 | - | 4 - 5 | 366 | 5685269 | 83 | |
U79773.1 | - | 4 - 5 | 398 | 83 | ||
U79774.1 | - | 4 - 5 | 398 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |