Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Per3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289877.1 | NP_001276806.1 | 1 - 2 | 559 | 118 | |
NM_001289878.1 | NP_001276807.1 | 1 - 2 | 559 | 118 | ||
NM_011067.3 | NP_035197.2 | 1 - 2 | 559 | 118 | ||
GenBank mRNA | AB013605.1 | - | 1 - 2 | 366 | 118 | |
AF050182.1 | - | 1 - 2 | 483 | 118 | ||
AJ007377.1 | - | 1 - 2 | 366 | 118 | ||
AK036345.1 | - | 1 - 2 | 476 | 118 | ||
AK166514.1 | - | 1 - 2 | 560 | 118 | ||
BC141032.1 | - | 1 - 2 | 157 | 9055644 | 118 | |
BC145178.1 | - | 1 - 2 | 157 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |