Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018790.3 | NP_061260.1 | 1 - 2 | 2011 | 71 | |
GenBank mRNA | AF162777.1 | - | 1 - 2 | 2002 | 71 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |