Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4e3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025829.4 | NP_080105.1 | 4 - 5 | 375 | 77 | |
GenBank mRNA | AK005054.2 | - | 4 - 5 | 376 | 77 | |
AY628329.1 | - | 4 - 5 | 356 | 77 | ||
BC027014.1 | - | 4 - 5 | 370 | 5369891 | 77 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |