Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008625.2 | NP_032651.2 | 7 - 8 | 1347 | 116 | |
GenBank mRNA | BC141338.1 | - | 7 - 8 | 1299 | 9055950 | 116 |
Z11974.1 | - | 7 - 8 | 1347 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |