Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK089402.1 | - | 4 - 5 | 404 | 69 | |
AK154742.1 | - | 4 - 5 | 404 | 69 | ||
AK155323.1 | - | 4 - 5 | 388 | 69 | ||
AK171362.1 | - | 4 - 5 | 401 | 69 | ||
BC061149.1 | - | 4 - 5 | 387 | 30305571 | 69 | |
U44795.1 | - | 4 - 5 | 393 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |