Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044371.2 | NP_001037836.1 | 3 - 4 | 472 | 96 | |
NM_001283011.1 | NP_001269940.1 | 3 - 4 | 472 | 96 | ||
NM_011233.3 | NP_035363.2 | 3 - 4 | 600 | 96 | ||
GenBank mRNA | AF085737.1 | - | 3 - 4 | 523 | 96 | |
AJ011923.1 | - | 2 - 3 | 373 | 96 | ||
AK134277.1 | - | 3 - 4 | 427 | 96 | ||
AK162016.1 | - | 3 - 4 | 459 | 96 | ||
AK167358.1 | - | 3 - 4 | 415 | 96 | ||
BC023939.1 | - | 3 - 4 | 477 | 96 | ||
BC066855.1 | - | 3 - 4 | 590 | 30473984 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |