Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rundc3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252347.1 | NP_001239276.1 | 2 - 3 | 494 | 89 | |
NM_016759.4 | NP_058039.1 | 2 - 3 | 494 | 89 | ||
GenBank mRNA | BC046319.1 | - | 2 - 3 | 394 | 6391042 | 89 |
U73941.1 | - | 2 - 3 | 221 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |