Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164733.1 | NP_001158205.1 | 4 - 5 | 551 | 92 | |
NM_001164734.1 | NP_001158206.1 | 4 - 5 | 561 | 92 | ||
NM_019939.2 | NP_064323.2 | 4 - 5 | 561 | 92 | ||
GenBank mRNA | AF161181.1 | - | 4 - 5 | 425 | 92 | |
AK144434.1 | - | 4 - 5 | 562 | 92 | ||
AK164549.1 | - | 4 - 5 | 552 | 92 | ||
BC042868.2 | - | 4 - 5 | 508 | 92 | ||
BC046421.1 | - | 4 - 5 | 508 | 92 | ||
BC069908.1 | - | 4 - 5 | 550 | 92 | ||
BC106140.1 | - | 4 - 5 | 508 | 92 | ||
BC138666.1 | - | 4 - 5 | 419 | 8861687 | 92 | |
BC145363.1 | - | 4 - 5 | 419 | 9053921 | 92 | |
BC145364.1 | - | 4 - 5 | 409 | 9053922 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |