Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK028796.1 | - | 7 - 8 | 1580 | 85 | |
AK156499.1 | - | 5 - 6 | 1539 | 85 | ||
AK157297.1 | - | 6 - 7 | 1449 | 85 | ||
AK165124.1 | - | 4 - 5 | 769 | 85 | ||
AK171128.1 | - | 7 - 8 | 1523 | 85 | ||
AK172551.1 | - | 6 - 7 | 1417 | 83 | ||
AK172573.1 | - | 6 - 7 | 1453 | 85 | ||
BC006759.1 | - | 6 - 7 | 1433 | 3709861 | 85 | |
U89992.2 | - | 1344 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |