Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tacc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040435.2 | NP_001035525.1 | 2 - 3 | 748 | 98 | |
GenBank mRNA | AF093542.1 | - | 2 - 3 | 249 | 98 | |
AF156934.1 | - | 2 - 3 | 299 | 98 | ||
AF203445.1 | - | 2 - 3 | 249 | 98 | ||
AF203446.1 | - | 2 - 3 | 249 | 98 | ||
AF247674.1 | - | 2 - 3 | 249 | 98 | ||
AK145556.1 | - | 2 - 3 | 309 | 98 | ||
AK160291.1 | - | 2 - 3 | 310 | 98 | ||
BC003252.1 | - | 2 - 3 | 284 | 3500413 | 98 | |
BC055071.1 | - | 2724 | 98 | |||
BC066184.1 | - | 2 - 3 | 284 | 30457053 | 98 | |
BC068314.1 | - | 2 - 3 | 284 | 30461783 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |