Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk32c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021302.3 | NP_067277.2 | 1 - 2 | 397 | 67 | |
GenBank mRNA | AB041542.1 | - | 1 - 2 | 377 | 67 | |
AK046439.1 | - | 1 - 2 | 375 | 67 | ||
AU080917.1 | - | 397 | 67 | |||
BC026457.1 | - | 1 - 2 | 382 | 4481218 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |